Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nr3c1
Entrez Gene ID
24413
Gene Name
nuclear receptor subfamily 3, group C, member 1
Gene Aliases
GR, Gcr, Grl
Chromosome Location
Chr.18: 31728373 - 32673988 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012576.2 NP_036708.2 1 - 2 1253 73
XM_008772065.1 XP_008770287.1 1 - 2 1254 73
XM_008772066.1 XP_008770288.1 2 - 3 1304 73
GenBank mRNA AB115420.1 - 1 - 2 1240 73
AF455050.1 - 1 - 2 1244 73
AY066016.2 - 1 - 2 1248 73
M14053.1 - 1 - 2 1311 73
Y00489.1 - 1 - 2 1243 73
Y12264.1 - 1 - 2 1265 73

View Full Product Details