The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012861.1 | NP_036993.1 | 4 - 5 | 503 | 76 |
GenBank mRNA | M76704.1 | - | 3 - 4 | 448 | 76 |
S61804.1 | - | 4 - 5 | 503 | 76 | |
X54862.1 | - | 4 - 5 | 529 | 76 |