Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ina
Entrez Gene ID
24503
Gene Name
internexin neuronal intermediate filament protein, alpha
Gene Aliases
Inexa, Intlaa, Nf66
Chromosome Location
Chr.1: 266781617 - 266794428 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019128.4 NP_062001.1 2 - 3 2480 87
GenBank mRNA X52017.1 - 2 - 3 1207 87

View Full Product Details