Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nptxr
Entrez Gene ID
81005
Gene Name
neuronal pentraxin receptor
Gene Aliases
Npr
Chromosome Location
Chr.7: 121011678 - 121029754 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030841.3 NP_110468.2 3 - 4 1195 65

View Full Product Details