The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130424.2 | NP_569108.2 | 7 - 8 | 786 | 76 |
XM_008768547.1 | XP_008766769.1 | 7 - 8 | 902 | 76 | |
XM_008768548.1 | XP_008766770.1 | 7 - 8 | 833 | 76 | |
XM_008768549.1 | XP_008766771.1 | 7 - 8 | 1015 | 76 | |
GenBank mRNA | AB073550.1 | - | 5 - 6 | 684 | 76 |
BC061712.1 | - | 7 - 8 | 786 | 76 |