Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chpf
Entrez Gene ID
316533
Gene Name
chondroitin polymerizing factor
Gene Aliases
D1bwg1363e
Chromosome Location
Chr.9: 82669199 - 82673898 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005906.1 NP_001005906.1 1 - 2 503 56
GenBank mRNA BC083617.1 - 1 - 2 503 56

View Full Product Details