Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nhej1
Entrez Gene ID
363251
Gene Name
nonhomologous end-joining factor 1
Gene Aliases
-
Chromosome Location
Chr.9: 82230230 - 82327923 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014217.1 NP_001014239.1 6 - 7 819 60
XM_006245240.1 XP_006245302.1 6 - 7 897 60
XM_006245241.2 XP_006245303.1 6 - 7 1473 60
XM_006245242.2 XP_006245304.1 6 - 7 1242 60
GenBank mRNA BC079033.1 - 6 - 7 819 60

View Full Product Details