The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011930.1 | NP_001011930.1 | 12 - 13 | 1318 | 64 |
XM_008758681.1 | XP_008756903.1 | 12 - 13 | 1320 | 64 | |
GenBank mRNA | BC083900.1 | - | 12 - 13 | 1318 | 64 |