Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ybx1
- Entrez Gene ID
-
500538
- Gene Name
- Y box binding protein 1
- Gene Aliases
-
Byb1, Cbfa, Dbpb, Ef1a, Msy1, Nsep1, Yb1
- Chromosome Location
-
Chr.5: 138319753 - 138336721 on Build Rnor_6.0
- Gene Symbol
- Ybx1-ps3
- Entrez Gene ID
-
29206
- Gene Name
- Y box protein 1 related, pseudogene 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 40380357 - 40381897 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038098.1 |
|
1 - 1 |
787 |
76 |
View Full Product Details