The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100922.1 | NP_001094392.1 | 9 - 10 | 1072 | 105 |
XM_006255219.1 | XP_006255281.1 | 9 - 10 | 1006 | 105 | |
GenBank mRNA | BC129128.1 | - | 9 - 10 | 1072 | 105 |
X57986.1 | - | 9 - 10 | 982 | 105 |