Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agpat2
Entrez Gene ID
311821
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.3: 4044741 - 4055384 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107821.2 NP_001101291.1 2 - 3 319 86

View Full Product Details