The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106216.1 | NP_001099686.1 | 1 - 1 | 1138 | 95 |
XM_006227988.2 | XP_006228050.1 | 2 - 2 | 1325 | 95 |