Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm5a
Entrez Gene ID
312678
Gene Name
lysine (K)-specific demethylase 5A
Gene Aliases
Jarid1a
Chromosome Location
Chr.4: 152892388 - 152972196 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277177.1 NP_001264106.1 7 - 8 1278 68
NM_001277178.1 NP_001264107.1 7 - 8 1278 68
GenBank mRNA BC099835.1 - 7 - 8 1021 68
BC169044.1 - 7 - 8 1278 68

View Full Product Details