Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmvk
Entrez Gene ID
310645
Gene Name
phosphomevalonate kinase
Gene Aliases
-
Chromosome Location
Chr.2: 188784329 - 188793885 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008352.1 NP_001008353.1 2 - 3 207 84
GenBank mRNA BC086349.1 - 2 - 3 207 84
FQ220630.1 - 2 - 3 257 84
FQ226470.1 - 2 - 3 337 84
FQ234368.1 - 2 - 3 337 84

View Full Product Details