Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Armcx6
Entrez Gene ID
363496
Gene Name
armadillo repeat containing, X-linked 6
Gene Aliases
-
Chromosome Location
Chr.X: 105565733 - 105573353 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257228.2 XP_006257290.1 1 - 2 178 78
XM_008773423.1 XP_008771645.1 4 - 5 1473 78
XM_008773424.1 XP_008771646.1 4 - 5 2475 78
XM_008773425.1 XP_008771647.1 1 - 2 178 78

View Full Product Details