The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013057.1 | NP_001013075.1 | 4 - 5 | 566 | 126 |
XM_006254183.2 | XP_006254245.1 | 4 - 5 | 578 | 126 | |
XM_006254184.2 | XP_006254246.1 | 4 - 5 | 557 | 126 | |
GenBank mRNA | BC078957.1 | - | 4 - 5 | 566 | 126 |