Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dusp27
Entrez Gene ID
498267
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
RGD1560598
Chromosome Location
Chr.13: 84302824 - 84332859 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191929.1 NP_001178858.1 1 - 2 108 148
XM_006250187.2 XP_006250249.1 3 - 4 308 148
XM_006250188.2 XP_006250250.1 3 - 4 304 148

View Full Product Details