The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008773.2 | NP_001008773.2 | 2 - 3 | 235 | 93 |
GenBank mRNA | BC093607.1 | - | 2 - 3 | 235 | 93 |