Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il18bp
Entrez Gene ID
84388
Gene Name
interleukin 18 binding protein
Gene Aliases
Igifbp
Chromosome Location
Chr.1: 167091521 - 167095718 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053374.2 NP_445826.2 1 - 2 151 113
XM_008759805.1 XP_008758027.1 1 - 2 2309 113
GenBank mRNA CB809932.1 - 1 - 2 162 113

View Full Product Details