Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl23
Entrez Gene ID
311114
Gene Name
kelch-like family member 23
Gene Aliases
RGD1307166
Chromosome Location
Chr.3: 56057043 - 56070614 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134504.1 NP_001127976.1 4 - 5 1768 95

View Full Product Details