The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039012.1 | NP_001034101.1 | 5 - 6 | 420 | 81 |
GenBank mRNA | BC100270.1 | - | 5 - 6 | 420 | 81 |