The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085353.1 | NP_001078822.1 | 11 - 12 | 1695 | 103 |
NM_199408.2 | NP_955440.1 | 11 - 12 | 1701 | 103 | |
GenBank mRNA | AY569010.1 | - | 11 - 12 | 1701 | 103 |
BC062391.1 | - | 11 - 12 | 1695 | 103 |