Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pate2
Entrez Gene ID
100361465
Gene Name
prostate and testis expressed 2
Gene Aliases
-
Chromosome Location
Chr.8: 36850324 - 36875241 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267781.1 NP_001254710.1 3 - 4 186 92
XM_006242781.2 XP_006242843.1 1 - 2 769 92
GenBank mRNA HQ687476.1 - 3 - 4 186 92

View Full Product Details