Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ran
Entrez Gene ID
84509
Gene Name
RAN, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.12: 31319556 - 31323758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053439.1 NP_445891.1 1 - 2 47 139
GenBank mRNA AF306457.1 - 1 - 2 47 139
FQ220799.1 - 1 - 2 75 139
FQ229761.1 - 1 - 2 38 139

View Full Product Details