The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106756.1 | NP_001100226.1 | 2 - 3 | 626 | 125 |
GenBank mRNA | FQ211881.1 | - | 2 - 3 | 341 | 125 |
FQ217020.1 | - | 2 - 3 | 342 | 125 | |
FQ223751.1 | - | 2 - 3 | 344 | 125 | |
FQ224251.1 | - | 2 - 3 | 341 | 125 | |
FQ224299.1 | - | 2 - 3 | 342 | 125 | |
FQ224538.1 | - | 2 - 3 | 355 | 125 |