Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exosc2
Entrez Gene ID
366017
Gene Name
exosome component 2
Gene Aliases
-
Chromosome Location
Chr.3: 10151734 - 10161997 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108952.1 NP_001102422.1 2 - 3 245 68
GenBank mRNA FQ227372.1 - 2 - 3 237 68

View Full Product Details