The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145401.1 | NP_001138873.1 | 12 - 12 | 2962 | 109 |
XM_008758921.1 | XP_008757143.1 | 12 - 12 | 3311 | 109 | |
XM_008758922.1 | XP_008757144.1 | 12 - 12 | 3284 | 109 | |
GenBank mRNA | BC161877.1 | - | 13 - 13 | 1869 | 109 |