Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Defa8
Entrez Gene ID
613224
Gene Name
defensin alpha 8
Gene Aliases
-
Chromosome Location
Chr.16: 75428675 - 75429568 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033077.1 NP_001028249.1 2 - 2 248 107
GenBank mRNA AY623752.1 - 2 - 2 248 107

View Full Product Details