Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STB4
Entrez Gene ID
855033
Gene Name
Gene Aliases
YMR019W
Chromosome Location
Chr.XIII: 312156 - 315005 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182515.1 NP_013732.1 1 - 1 2751 85

View Full Product Details