Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OXT
Entrez Gene ID
100152272
Gene Name
oxytocin/neurophysin I prepropeptide
Gene Aliases
oxytocin
Chromosome Location
Chr.17: 32605586 - 32606449 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167589.1 NP_001161061.1 2 - 3 321 81

View Full Product Details