Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HINT2
Entrez Gene ID
100512882
Gene Name
histidine triad nucleotide binding protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 236532316 - 236535263 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244325.1 NP_001231254.1 2 - 3 240 107
XM_005660206.3 XP_005660263.1 - 708 107
GenBank mRNA AK346311.1 - 2 - 3 237 107
AK392440.1 - 2 - 3 227 107
AY609781.1 - 2 - 3 227 107
EW338246.2 - 2 - 3 242 107

View Full Product Details