Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
med10
Entrez Gene ID
448288
Gene Name
mediator complex subunit 10
Gene Aliases
-
Chromosome Location
Chr.6: 60241796 - 60256598 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004914.1 NP_001004914.1 2 - 3 324 95
GenBank mRNA BC075360.1 - 2 - 3 324 95

View Full Product Details