Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp2f2
Entrez Gene ID
496408
Gene Name
Cyp2f2 protein
Gene Aliases
-
Chromosome Location
Chr.8: 57597195 - 57616616 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010999.1 NP_001010999.1 7 - 8 1188 75
GenBank mRNA BC076922.1 - 7 - 8 1188 75

View Full Product Details