Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ela1
Entrez Gene ID
595061
Gene Name
ela1 protein
Gene Aliases
-
Chromosome Location
Chr.2: 139128578 - 139135683 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030498.1 NP_001025669.1 7 - 8 788 116
GenBank mRNA BC092553.1 - 7 - 8 788 116

View Full Product Details