Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
akr1c2
Entrez Gene ID
779650
Gene Name
aldo-keto reductase family 1, member C2
Gene Aliases
akr1c2-a, akr1c2-b, babp, dd2, ddh2, hakrd, hbab, mcdr2, srxy8
Chromosome Location
Chr.3: 1485005 - 1491851 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078736.1 NP_001072204.1 2 - 3 277 76
GenBank mRNA BC118758.1 - 2 - 3 277 76
CU025147.1 - 2 - 3 277 76

View Full Product Details