Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gm2a
Entrez Gene ID
548463
Gene Name
GM2 ganglioside activator
Gene Aliases
-
Chromosome Location
Chr.3: 11055763 - 11069457 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012958955.2 XP_012814409.1 4 - 4 1780 97
GenBank mRNA BC089682.1 - 4 - 4 1749 97

View Full Product Details