Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
atg9
Entrez Gene ID
732722
Gene Name
autophagy protein 9
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112351.1 NP_001105821.1 - 1839 72
GenBank mRNA AM085516.1 - - 1839 72

View Full Product Details