Gene Transcripts

Gene Symbol
Dlx6os2
Entrez Gene ID
13397
Gene Name
distal-less homeobox 6, opposite strand 2
Gene Aliases
Dlx6as, Dlx6as2
Chromosome Location
Chr. 6: 6863797 - 6865150 on Build GRCm38
UniGene ID
Mm.482625
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002839.2
1 684
GenBank mRNA AF022079.1 - LIT3325
Dlx6as
1 682

View Full Product Details