Gene Transcripts
- Gene Symbol
- FTH1P3
- Entrez Gene ID
- 2498
- Gene Name
- ferritin heavy chain 1 pseudogene 3
- Gene Aliases
- FTHL3, FTHL3P
- Chromosome Location
- Chr. 2: 27392623 - 27393576 on Build GRCh38
- UniGene ID
- Hs.658438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002201.1 |
|
1 |
520 |
|
GenBank mRNA |
BC073777.1 |
- |
1 |
531 |
|
View Full Product Details