Gene Transcripts
- Gene Symbol
- LINC01554
- Entrez Gene ID
- 202299
- Gene Name
- long intergenic non-protein coding RNA 1554
- Gene Aliases
- C5orf27, FIS
- Chromosome Location
- Chr. 5: 95852232 - 95860133 on Build GRCh38
- UniGene ID
- Hs.8373
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026936.1 |
|
3 |
1132 |
|
GenBank mRNA |
AK096140.1 |
- |
4 |
1031 |
|
AY168789.1 |
- |
4 |
1011 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000620167.1 |
- |
|
658 |
|
Chr. - - [-] |
View Full Product Details