Gene Transcripts
- Gene Symbol
- SNORA74B
- Entrez Gene ID
- 677841
- Gene Name
- small nucleolar RNA, H/ACA box 74B
- Gene Aliases
- U19-2
- Chromosome Location
- Chr. 5: 173020726 - 173020929 on Build GRCh38
- UniGene ID
- Hs.692720
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002988.1 |
|
1 |
78 |
|
GenBank mRNA |
AM055743.1 |
- |
1 |
78 |
|
View Full Product Details