Gene Transcripts
- Gene Symbol
- SPRED3
- Entrez Gene ID
- 399473
- Gene Name
- sprouty related EVH1 domain containing 3
- Gene Aliases
- Eve-3, spred-3
- Chromosome Location
- Chr. 19: 38390061 - 38399884 on Build GRCh38
- UniGene ID
- Hs.196011
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
W79238.1 |
- |
|
272 |
|
Chr. - - [-] |
GenBankmRNA |
AF086394.1 |
- |
1 |
272 |
|
Chr. 1 - 38912988 - 38913527 [-] |
View Full Product Details