Gene Transcripts
- Gene Symbol
- C1orf61
- Entrez Gene ID
- 10485
- Gene Name
- chromosome 1 open reading frame 61
- Gene Aliases
- CROC4
- Chromosome Location
- Chr. 1: 156404252 - 156456649 on Build GRCh38
- UniGene ID
- Hs.380027
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095622.1 |
- |
1 |
1486 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BF530499.1 |
- |
1 |
241 |
|
Chr. 1 - 156411202 - 156411639 [-] |
View Full Product Details