Gene Transcripts

Gene Symbol
C1orf61
Entrez Gene ID
10485
Gene Name
chromosome 1 open reading frame 61
Gene Aliases
CROC4
Chromosome Location
Chr. 1: 156404252 - 156456649 on Build GRCh38
UniGene ID
Hs.380027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK095622.1 - 1 2612

View Full Product Details