Gene Transcripts
- Gene Symbol
- Zscan30
- Entrez Gene ID
- 328918
- Gene Name
- zinc finger and SCAN domain containing 30
- Gene Aliases
- C230097I24Rik, Gm754, Zfp397os
- Chromosome Location
- Chr. 18: 23932540 - 23987348 on Build GRCm38
- UniGene ID
- Mm.261685
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053535.1 |
- |
1 |
1938 |
|
View Full Product Details