Gene Transcripts
- Gene Symbol
- Vmn1r84
- Entrez Gene ID
- 171267
- Gene Name
- vomeronasal 1 receptor 84
- Gene Aliases
- V1rg11
- Chromosome Location
- Chr. 7: 12361808 - 12362764 on Build GRCm38
- UniGene ID
- Mm.434341
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134233.1 |
NP_598994.1
|
1 |
793 |
|
View Full Product Details