Gene Transcripts

Gene Symbol
Zdhhc5
Entrez Gene ID
228136
Gene Name
zinc finger, DHHC domain containing 5
Gene Aliases
1110032A17Rik, AI451382, Zisp
Chromosome Location
Chr. 2: 84687920 - 84715164 on Build GRCm38
UniGene ID
Mm.288508
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144887.4 NP_659136.1 11 2881
GenBank mRNA AK028737.1 - 8 1447
AK075641.1 - 1 228
AK082513.1 - 11 2806
AK145086.1 - 11 2809
AK173251.1 - 11 2836
AY894891.1 - 11 2873
BC020051.1 - 10 1851 3987696
BC027047.1 - 7 1115
BC065155.1 - 11 2751 5706087

View Full Product Details