Gene Transcripts

Gene Symbol
PSMD1
Entrez Gene ID
5707
Gene Name
proteasome 26S subunit, non-ATPase 1
Gene Aliases
P112, Rpn2, S1
Chromosome Location
Chr. 2: 231056864 - 231172827 on Build GRCh38
UniGene ID
Hs.3887
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191037.1 NP_001177966.1 11, 12 1394
NM_002807.3 NP_002798.2 11, 12 1394
NR_034059.1 10, 11 1224
XM_017004517.1 XP_016860006.1 11, 12 1394
GenBank mRNA AK307415.1 - 10, 11 1221
AK307513.1 - 9, 10 1096
AK312528.1 - 11, 12 1351
BC001053.2 - 11, 12 1351
BC014013.2 - 11, 12 1363
BC020223.1 - 11, 12 1341
BC032445.1 - 11, 12 1366
BC039845.1 - 11, 12 1317
BC047897.1 - 11, 12 1316
BC064398.1 - 11, 12 1365
BC073833.1 - 11, 12 1318
BC094720.1 - 11, 12 1380 4826340
BC112344.1 - 11, 12 1355 40008454
BC114434.1 - 11, 12 1355 40008444
D44466.1 - 11, 12 1312
KU921434.1 - 4, 5 351

View Full Product Details