Gene Transcripts

Gene Symbol
Nynrin
Entrez Gene ID
277154
Gene Name
NYN domain and retroviral integrase containing
Gene Aliases
AA409373, C80530, mKIAA1305
Chromosome Location
Chr. 14: 55854115 - 55874736 on Build GRCm38
UniGene ID
Mm.319347
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040072.1 NP_001035161.1 8 3602
GenBank mRNA AK220380.1 - 6 1451
BC057379.1 - 2 435
BC151043.1 - 9 3624 9087956

View Full Product Details