Gene Transcripts

Gene Symbol
Zscan30
Entrez Gene ID
328918
Gene Name
zinc finger and SCAN domain containing 30
Gene Aliases
C230097I24Rik, Gm754, Zfp397os
Chromosome Location
Chr. 18: 23932540 - 23987348 on Build GRCm38
UniGene ID
Mm.261685
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK049082.1 - 3 697
BC117067.1 - 2 478 40126386
BC117069.1 - 2 478 40126388

View Full Product Details